Invitrogen
Manufacturer Code:PA5114804
Catalog # PIPA5114804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ABCG4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6430517O04Rik ABCG4 ATP binding cassette subfamily G member 4 ATP-binding cassette sub-family G member 4 ATP-binding cassette subfamily G (WHITE) member 4 ATP-binding cassette sub-family G (WHITE) member 4 putative ABC transporter WHITE2; ATP-binding cassette sub-family G member 4; ATP-binding cassette, sub-family G (WHITE), member 4; putative ABC transporter
Gene Aliases: ABCG4; WHITE2
UniProt ID: (Human) Q9H172
Entrez Gene ID: (Human) 64137
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.