Proteintech
Manufacturer Code:142691AP150UL
Catalog # 50-172-9330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ABCG4 Fusion Protein Ag5587 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABCG4 WHITE2; ATP-binding cassette sub-family G member 4; ATP-binding cassette, sub-family G (WHITE), member 4; putative ABC transporter
Gene Aliases: 6430517O04Rik; ABCG4; WHITE2
UniProt ID: (Human) Q9H172
Entrez Gene ID: (Human) 64137, (Mouse) 192663, (Rat) 300664
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.