R&D Systems
Manufacturer Code:MAB995100
Catalog # MAB995100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5D3 |
Immunogen | 3T3 cells transduced with human ABCG2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ABC transporter; ABC transporter ABC15 ABCP ABCPMGC102821 ATP-binding cassette sub-family G member 2 ATP-binding cassette transporter G2 ATP-binding cassette sub-family G (WHITE) member 2 BCRP BCRP1 BCRPMRX BMDP Breast cancer resistance protein CD338 CD338 antigen CDw338 EST157481 mitoxantrone resistance protein Mitoxantrone resistance-associated protein MXR MXRMXR1 placenta specific MDR protein Placenta-specific ATP-binding cassette transporter; ATP-binding cassette sub-family G member 2; ATP-binding cassette transporter G2; ATP-binding cassette, sub-family G (WHITE), member 2 (Junior blood group); Breast cancer resistance protein; Broad substrate specificity ATP-binding cassette transporter ABCG2; CD338; CDw338; Mitoxantrone resistance-associated protein; multi drug resistance efflux transport ATP-binding cassette sub-family G (WHITE) member 2; placenta specific MDR protein; Placenta-specific ATP-binding cassette transporter; Urate exporter
Gene Aliases: ABC15; ABCG2; ABCP; BCRP; BCRP1; BMDP; CD338; CDw338; EST157481; GOUT1; MRX; MXR; MXR-1; MXR1; UAQTL1
UniProt ID: (Human) Q9UNQ0
Entrez Gene ID: (Human) 9429
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.