Abnova
Manufacturer Code:MAB5284
Catalog # 89-112-204
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | BXP-21 |
Immunogen | Recombinant protein corresponding to amino acids 271-396 of human ABCG2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant ABCG2.
For Research Use Only
Protein Aliases: ABC transporter; ABC15; ABCP; ATP-binding cassette sub-family G member 2; ATP-binding cassette transporter G2; ATP-binding cassette, sub-family G (WHITE), member 2 (Junior blood group); BCRP; BCRP1; BMDP; Breast cancer resistance protein; Broad substrate specificity ATP-binding cassette transporter ABCG2; CD338; CDw338; EST157481; MGC102821; Mitoxantrone resistance-associated protein; MRX; multi drug resistance efflux transport ATP-binding cassette sub-family G (WHITE) member 2; MXR; MXR1; placenta specific MDR protein; Placenta-specific ATP-binding cassette transporter; Urate exporter
Gene Aliases: ABC15; ABCG2; ABCP; BCRP; BCRP1; BMDP; CD338; CDw338; EST157481; GOUT1; MRX; MXR; MXR-1; MXR1; UAQTL1
UniProt ID: (Human) Q9UNQ0
Entrez Gene ID: (Human) 9429
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.