Invitrogen
Manufacturer Code:PA595071
Catalog # PIPA595071
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human ABCG1 identical to the related rat and mouse sequences. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC transporter 8; ATP-binding cassette 8; ATP-binding cassette sub-family G member 1; ATP-binding cassette sub-family G member 1 ATP-binding cassette transporter 8 White protein homolog ABCG1 ABC8 WHT1; ATP-binding cassette transporter 8; ATP-binding cassette transporter member 1 of subfamily G; ATP-binding cassette, sub-family G (WHITE), member 1; ATP-binding cassette, subfamily G, member 1; homolog of Drosophila white; White protein homolog; white protein homolog (ATP-binding cassette transporter 8)
Gene Aliases: ABC8; ABCG1; AW413978; White; WHITE1; WHT1
UniProt ID: (Human) P45844, (Mouse) Q64343
Entrez Gene ID: (Human) 9619, (Rat) 85264, (Mouse) 11307
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.