Novus Biologicals
Manufacturer Code:NB400132PEATT594
Catalog # N40132PE594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal region of human ABCG1 (between residues 300-400). [UniProt No. P45844] |
Conjugate | PE/ATTO 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Rabbit Polyclonal Antibody
Protein Aliases: ABC transporter 8; ABC transporter 8 ABC8ATP-binding cassette sub-family G member 1 ATP-binding cassette transporter 8 ATP-binding cassette sub-family G (WHITE) member 1 homolog of Drosophila white MGC34313 White protein homolog white protein homolog (ATP-binding cassette transporter 8) 10ATP-binding cassette transporter member 1 of subfamily G WHITE1 WHT1; ATP-binding cassette sub-family G member 1; ATP-binding cassette transporter 8; ATP-binding cassette transporter member 1 of subfamily G; ATP-binding cassette, sub-family G (WHITE), member 1; homolog of Drosophila white; White protein homolog; white protein homolog (ATP-binding cassette transporter 8)
Gene Aliases: ABC8; ABCG1; WHITE1; WHT1
UniProt ID: (Human) P45844
Entrez Gene ID: (Human) 9619
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.