Abnova
Manufacturer Code:H00009619K
Catalog # 89-203-185
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ABCG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ABCG1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: ABC transporter 8; ABC8 MGC34313 WHITE1; ATP-binding cassette sub-family G member 1; ATP-binding cassette transporter 8; ATP-binding cassette transporter member 1 of subfamily G; ATP-binding cassette, sub-family G (WHITE), member 1; homolog of Drosophila white; White protein homolog; white protein homolog (ATP-binding cassette transporter 8)
Gene Aliases: ABC8; ABCG1; WHITE1; WHT1
UniProt ID: (Human) P45844
Entrez Gene ID: (Human) 9619
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.