OriGene
Manufacturer Code:TA811212
Catalog # 50-168-3909
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5D5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-235 of human ABCF2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ABC-type transport protein; ABC28 EST133090 HUSSY-18 HUSSY18; abcf2; ATP-binding cassette sub-family F member 2; ATP-binding cassette, sub-family F (GCN20), member 2; developmentally regulated repeat element-containing transcript 3; Iron-inhibited ABC transporter 2
Gene Aliases: 0710005O05Rik; ABC28; ABCF2; Drr3; E430001O06; EST133090; HUSSY-18; HUSSY18
UniProt ID: (Human) Q9UG63, (Mouse) Q99LE6
Entrez Gene ID: (Human) 10061, (Mouse) 27407
Molecular Function: ATP-binding cassette (ABC) transporter RNA binding protein hydrolase nucleic acid binding translation elongation factor translation factor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.