Invitrogen
Manufacturer Code:PA554143
Catalog # PIPA554143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ABCF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC-type transport protein; ABC28 DKFZp586K1823 EST133090 HUSSY-18 HUSSY18 M-ABC1; abcf2; ATP-binding cassette sub-family F member 2; ATP-binding cassette, sub-family F (GCN20), member 2; developmentally regulated repeat element-containing transcript 3; Iron-inhibited ABC transporter 2
Gene Aliases: 0710005O05Rik; ABC28; ABCF2; Drr3; E430001O06; EST133090; HUSSY-18; HUSSY18
UniProt ID: (Human) Q9UG63, (Mouse) Q99LE6
Entrez Gene ID: (Human) 10061, (Mouse) 27407, (Rat) 311959
Molecular Function:
ATP-binding cassette (ABC) transporter
RNA binding protein
hydrolase
nucleic acid binding
translation elongation factor
translation factor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.