Novus Biologicals
Manufacturer Code:NB400115
Catalog # NB400115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A fusion protein containing amino acids 1-102 of the ABCF2 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC-type transport protein; ABC28 ABC-type transport protein ATP-binding cassette sub-family F member 2 ATP-binding cassette sub-family F (GCN20) member 2 DKFZp586K1823 EC 3.6.3 EC 3.6.3.17 EST133090 HUSSY18 HUSSY-18 Iron inhibited ABC transporter 2 Iron-inhibited ABC transporter 2 M-ABC1; ATP-binding cassette sub-family F member 2; ATP-binding cassette, sub-family F (GCN20), member 2; Iron-inhibited ABC transporter 2
Gene Aliases: ABC28; ABCF2; EST133090; HUSSY-18; HUSSY18
UniProt ID: (Human) Q9UG63
Entrez Gene ID: (Human) 10061
Molecular Function: ATP-binding cassette (ABC) transporter RNA binding protein hydrolase nucleic acid binding translation elongation factor translation factor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.