Novus Biologicals
Manufacturer Code:NBP267698
Catalog # NBP267698
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SN07-12 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: ABC27 ABC50TNFalpha-inducible ATP-binding protein ATP-binding cassette 50 ATP-binding cassette 50 (TNF-alpha stimulated) ATP-binding cassette sub-family F member 1 ATP-binding cassette sub-family F (GCN20) member 1 EC 2.7.7.6 EC 3.6.3.17 EST123147 TNF-alpha-stimulated ABC protein; ATP-binding cassette 50; ATP-binding cassette 50 (TNF-alpha stimulated); ATP-binding cassette sub-family F member 1; ATP-binding cassette, sub-family F (GCN20), member 1; TNF-alpha-stimulated ABC protein; TNFalpha-inducible ATP-binding protein
Gene Aliases: ABC27; ABC50; ABCF1
UniProt ID: (Human) Q8NE71
Entrez Gene ID: (Human) 23
Molecular Function: ATP-binding cassette (ABC) transporter RNA binding protein hydrolase nucleic acid binding translation elongation factor translation factor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.