Invitrogen
Manufacturer Code:PA599218
Catalog # PIPA599218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ABCF1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC27 Abc50 Abcf1 ATP binding cassette subfamily F member 1 ATP-binding cassette 50 ATP-binding cassette 50 (TNF-alpha stimulated) ATP-binding cassette sub-family F member 1 ATP-binding cassette subfamily F (GCN20) member 1 ATP-binding cassette sub-family F (GCN20) member 1 AU041969 D17Wsu166e GCN20 TNFalpha-inducible ATP-binding protein TNF-alpha-stimulated ABC protein; ATP-binding cassette 50; ATP-binding cassette 50 (TNF-alpha stimulated); ATP-binding cassette sub-family F member 1; ATP-binding cassette, sub-family F (GCN20), member 1; TNF-alpha-stimulated ABC protein; TNFalpha-inducible ATP-binding protein
Gene Aliases: ABC27; ABC50; ABCF1; AU041969; D17Wsu166e; GCN20
UniProt ID: (Human) Q8NE71, (Mouse) Q6P542, (Rat) Q6MG08
Entrez Gene ID: (Human) 23, (Mouse) 224742, (Rat) 85493
Molecular Function:
ATP-binding cassette (ABC) transporter
RNA binding protein
hydrolase
nucleic acid binding
translation elongation factor
translation factor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.