Invitrogen
Manufacturer Code:PA116797
Catalog # APPA6797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Fusion protein containing residues 1-119 of human ABCE1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2'-5'-oligoadenylate-binding protein; ABC 38 ABC38 ABCE 1 ATP Binding Cassette Sub Family E Member 1 OABP Ribonuclease 4 Inhibitor RLI RNase L Inhibitor RNS4I RNase 4 inhibitor; ATP-binding cassette sub-family E member 1; ATP-binding cassette, sub-family E (OABP), member 1; HuHP68; Ribonuclease 4 inhibitor; ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent) inhibitor; RNase L inhibitor; RNase L inhibitor 1; RNS4I
Gene Aliases: ABC38; ABCE1; OABP; OK/SW-cl.40; RLI; RLI1; RNASEL1; RNASELI; RNS4I
UniProt ID: (Human) P61221
Entrez Gene ID: (Human) 6059
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.