Invitrogen
Manufacturer Code:PA5109352
Catalog # PIPA5109352
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 100-280 of human ABCD4 (NP_005041.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 69 kDa peroxisomal ABC-transporter; ABC41 ABCD4 ATP-binding cassette sub-family D member 4 EST352188 MAHCJ P69r P70R P79R Peroxisomal membrane protein 1-like Peroxisomal membrane protein 69 PMP69 PMP70-related protein PXMP1L PXMP1-L; ATP-binding cassette sub-family D member 4; ATP-binding cassette, sub-family D (ALD), member 4; Lysosomal cobalamin transporter ABCD4; P70R; Peroxisomal membrane protein 1-like; Peroxisomal membrane protein 69; PMP69; PMP70-related protein; PXMP1-L
Gene Aliases: ABC41; ABCD4; EST352188; MAHCJ; P70R; P79R; PMP69; PXMP1L
UniProt ID: (Human) O14678
Entrez Gene ID: (Human) 5826
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.