Novus Biologicals
Manufacturer Code:NBP120872
Catalog # NBP120872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-RDDIDNPDQRISQD corresponding to internal region according to NP_005041.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 69 kDa peroxisomal ABC-transporter; ATP-binding cassette sub-family D member 4; ATP-binding cassette sub-family D member 4 ATP-binding cassette sub-family D (ALD) member 4 EST352188 P70R69 kDa peroxisomal ABC-transporter Peroxisomal membrane protein 1-like Peroxisomal membrane protein 69 PMP69P79R PMP70-related protein PXMP1-L PXMP1LABC41; ATP-binding cassette, sub-family D (ALD), member 4; Lysosomal cobalamin transporter ABCD4; P70R; Peroxisomal membrane protein 1-like; Peroxisomal membrane protein 69; PMP69; PMP70-related protein; PXMP1-L
Gene Aliases: ABC41; ABCD4; EST352188; MAHCJ; P70R; P79R; PMP69; PXMP1L
UniProt ID: (Human) O14678
Entrez Gene ID: (Human) 5826
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.