Proteintech
Manufacturer Code:666971IG150UL
Catalog # 50-173-7113
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G7G5 |
Immunogen | ABCD3 Fusion Protein Ag24760 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 70 kDa peroxisomal membrane protein; ABC43 ABCD3 PMP70 PXMP1; ATP-binding cassette sub-family D member 3; ATP-binding cassette, sub-family D (ALD), member 3; dJ824O18.1 (ATP-binding cassette, sub-family D (ALD), member 3 (PMP70, PXMP1)); peroxisomal membrane protein 1 (70kD, Zellweger syndrome); Peroxisomal membrane protein-1 (70kD); PMP70
Gene Aliases: ABC43; ABCD3; CBAS5; PMP70; PXMP1; ZWS2
UniProt ID: (Human) P28288
Entrez Gene ID: (Human) 5825
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.