Invitrogen
Manufacturer Code:PA526632
Catalog # PIPA526632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 269-298 amino acids from the central region of human ABCD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC39 ALDL1 ALDR ALDRP hALDR ATP-binding cassette sub-family D member 2 adrenoleukodystrophy-like 1 adrenoleukodystrophy-related protein; adrenoleukodystrophy related; Adrenoleukodystrophy-like 1; Adrenoleukodystrophy-related protein; ATP-binding cassette sub-family D member 2; ATP-binding cassette, sub-family D (ALD), member 2; hALDR
Gene Aliases: ABC39; ABCD2; ALD1; ALDL1; ALDR; ALDRP; hALDR
UniProt ID: (Human) Q9UBJ2, (Mouse) Q61285
Entrez Gene ID: (Human) 225, (Mouse) 26874
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.