Novus Biologicals
Manufacturer Code:NBP292604100UL
Catalog # NB027078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 340-500 of human ABCD1 (NP_000024.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adrenoleukodystrophy protein; Adrenoleukodystrophy protein ALDATP-binding cassette sub-family D member 1 ALDPadrenoleukodystrophy AMNABC42 ATP-binding cassette sub-family D (ALD) member 1; ALDP; ATP-binding cassette sub-family D member 1; ATP-binding cassette, sub-family D (ALD), member 1
Gene Aliases: ABC42; ABCD1; ALD; ALDP; AMN
UniProt ID: (Human) P33897
Entrez Gene ID: (Human) 215
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.