Abnova
Manufacturer Code:PAB25128
Catalog # 89-126-801
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to ABCD1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ABCD1.
For Research Use Only
Protein Aliases: ABC42; Adrenoleukodystrophy protein; ALD; ALDP; AMN; ATP-binding cassette sub-family D member 1; ATP-binding cassette, sub-family D (ALD), member 1
Gene Aliases: ABC42; ABCD1; ALD; ALDP; AMN
UniProt ID: (Human) P33897
Entrez Gene ID: (Human) 215
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.