Proteintech
Manufacturer Code:111591AP150UL
Catalog # 50-172-6925
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ABCD1 Fusion Protein Ag1635 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC42 ABCD1 Adrenoleukodystrophy protein ALD ALDP AMN; Adrenoleukodystrophy protein; ALDP; ATP-binding cassette sub-family D member 1; ATP-binding cassette, sub-family D (ALD), member 1; ATP-binding cassette, sub-family D, member 1; X-linked adrenoleukodystrophy (ALD) gene homolog
Gene Aliases: ABC42; ABCD1; ALD; Aldgh; ALDP; AMN; RGD1562128
UniProt ID: (Human) P33897, (Mouse) P48410, (Rat) D3ZHR2
Entrez Gene ID: (Human) 215, (Mouse) 11666, (Rat) 363516
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.