Novus Biologicals
Manufacturer Code:NBP222403AF488
Catalog # NB222403488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S319A-14 |
Immunogen | Fusion protein amino acids 1505-1546 (SSIVDAGLVLVFSEGILVECDTGPNLLQHKNGLFSTLVMTNK cytoplasmic C-terminus) of mouse SUR2A |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ABC37 ATP-binding cassette sub-family C (CFTR/MRP) member 9 CMD1OATP-binding cassette transporter sub-family C member 9 EC 3.6.3.44 FLJ36852 Sulfonylurea receptor 2 sulfonylurea receptor 2A SUR2ATP-binding cassette sub-family C member 9; ATP-binding cassette sub-family C member 9; ATP-binding cassette transporter sub-family C member 9; ATP-binding cassette, sub-family C (CFTR/MRP), member 9; Sulfonylurea receptor 2
Gene Aliases: ABC37; ABCC9; ATFB12; CANTU; CMD1O; SUR2
UniProt ID: (Human) O60706
Entrez Gene ID: (Human) 10060
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.