Invitrogen
Manufacturer Code:PA542398
Catalog # PIPA542398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ABCC9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-binding cassette sub-family C (CFTR/MRP) member 9 ABC37 CMD1O FLJ36852 SUR2 ATFB12; ATP-binding cassette sub-family C member 9; ATP-binding cassette transporter sub-family C member 9; ATP-binding cassette, sub-family C (CFTR/MRP), member 9; cardiac ventricle sulfonyl urea receptor; membrane receptor protein; Sulfonylurea receptor 2; sulfonylurea receptor subunit 2; sulfonylurea-binding protein 2; sulphonylurea receptor 2; sulphonylurea receptor 2A; sulphonylurea receptor 2B; SUR2; SUR2A; SUR2B
Gene Aliases: ABC37; ABCC9; AI414027; AI449286; ATFB12; CANTU; CMD1O; si:dkey-183c2.3; SUR2; SUR2A; SUR2B
UniProt ID: (Rabbit) P82451, (Human) O60706, (Rat) Q63563, (Mouse) P70170
Entrez Gene ID: (Goat) 102187656, (Dog) 486638, (Rabbit) 100008700, (Human) 10060, (Horse) 100064351, (Rat) 25560, (Guinea pig) 100720522, (Mouse) 20928, (Zebrafish) 561160
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.