Novus Biologicals
Manufacturer Code:NBP159350
Catalog # NBP159350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ABCC9(ATP-binding cassette sub-family C (CFTR/MRP) member 9) The peptide sequence was selected from the middle region of ABCC9 (NP_005682). Peptide sequence AVVTEGGENFSVGQRQLFCLARAFVRKSSILIMDEATASIDMATENILQK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC37 ATP-binding cassette sub-family C (CFTR/MRP) member 9 CMD1OATP-binding cassette transporter sub-family C member 9 EC 3.6.3.44 FLJ36852 Sulfonylurea receptor 2 sulfonylurea receptor 2A SUR2ATP-binding cassette sub-family C member 9; ATP-binding cassette sub-family C member 9; ATP-binding cassette transporter sub-family C member 9; ATP-binding cassette, sub-family C (CFTR/MRP), member 9; Sulfonylurea receptor 2
Gene Aliases: ABC37; ABCC9; ATFB12; CANTU; CMD1O; SUR2
UniProt ID: (Human) O60706
Entrez Gene ID: (Human) 10060
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.