Abnova
Manufacturer Code:H00000368A01
Catalog # 89-104-099
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 831 a.a. to 930 a.a) partial recombinant protein with GST tag.,ABCC6 (NP_001162 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant ABCC6.
For Research Use Only
Protein Aliases: ABC34; Anthracycline resistance-associated protein; ARA; ATP-binding cassette sub-family C member 6; ATP-binding cassette, sub-family C (CFTR/MRP), member 6; EST349056; MLP1; MOAT-E; MOATE; MRP6; Multi-specific organic anion transporter E; Multidrug resistance-associated protein 6; PXE; PXE1
Gene Aliases: ABC34; ABCC6; ARA; EST349056; GACI2; MLP1; MOAT-E; MOATE; MRP6; PXE; PXE1; URG7
UniProt ID: (Human) O95255
Entrez Gene ID: (Human) 368
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.