Proteintech
Manufacturer Code:278481AP150UL
Catalog # 50-173-5775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ABCC6 Fusion Protein Ag18640 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC34 ABCC6 ARA EST349056 MLP1 MOAT E MOATE MRP6 PXE PXE1; Anthracycline resistance-associated protein; ATP-binding cassette sub-family C member 6; ATP-binding cassette, sub-family C (CFTR/MRP), member 6; ATP-binding cassette, sub-family C, member 6; MOAT-E; Multi-specific organic anion transporter E; Multidrug resistance-associated protein 6; multidrug resistance-associated protein-6
Gene Aliases: ABC34; Abcc1b; ABCC6; ARA; DCC; dyscalc; Dyscalc1; EST349056; GACI2; MLP1; MOAT-E; MOATE; MRP6; PXE; PXE1; URG7
UniProt ID: (Human) O95255, (Mouse) Q9R1S7
Entrez Gene ID: (Human) 368, (Mouse) 27421
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.