OriGene
Manufacturer Code:TA808370
Catalog # 50-168-2085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6C6 |
Immunogen | Full length human recombit protein of human ABCC5 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ABC33 EST277145 MOAT-C MOATC MRP5 SMRP pABC11; ATP-binding cassette sub-family C member 5; ATP-binding cassette, sub-family C (CFTR/MRP), member 5; canalicular multispecific organic anion transporter C; MOAT-C; Multi-specific organic anion transporter C; Multidrug resistance-associated protein 5; pABC11; SMRP
Gene Aliases: ABC33; ABCC5; EST277145; MOAT-C; MOATC; MRP5; pABC11; SMRP
UniProt ID: (Human) O15440
Entrez Gene ID: (Human) 10057
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.