Abnova
Manufacturer Code:MAB6675
Catalog # 89-112-999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | M9II-3 |
Immunogen | Recombinant protein corresponding to amino acids 690-734 of human ABCC12. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rat monoclonal antibody raised against partial recombinant ABCC12.
For Research Use Only
Protein Aliases: ATP-binding cassette sub-family C member 12; ATP-binding cassette transporter sub-family C member 12; ATP-binding cassette, sub-family C (CFTR/MRP), member 12; MGC27071; MRP9; Multidrug resistance-associated protein 9
Gene Aliases: ABCC12; MRP9
UniProt ID: (Human) Q96J65
Entrez Gene ID: (Human) 94160
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.