Abnova
Manufacturer Code:PAB31498
Catalog # 89-998-481
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human ABCC12. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-binding cassette sub-family C member 12; ATP-binding cassette transporter sub-family C member 12; ATP-binding cassette, sub-family C (CFTR/MRP), member 12; MGC27071; MRP9; Multidrug resistance-associated protein 9
Gene Aliases: ABCC12; MRP9
UniProt ID: (Human) Q96J65
Entrez Gene ID: (Human) 94160
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.