Invitrogen
Manufacturer Code:MA521478
Catalog # PIMA521478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F4 |
Immunogen | ABCB9 (NP&lowbar062571 482 a.a. &bsim 580 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ABC transporter 9 protein; ABCB9 ATP-binding cassette sub-family B (MDR/TAP) member 9 TAPL KIAA1520 EST122234; ATP-binding cassette sub-family B member 9; ATP-binding cassette transporter 9; ATP-binding cassette, sub-family B (MDR/TAP), member 9; TAP-like protein; TAPL
Gene Aliases: ABCB9; EST122234; KIAA1520; TAPL
UniProt ID: (Human) Q5W9G7
Entrez Gene ID: (Human) 23457
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.