Novus Biologicals
Manufacturer Code:NBP159553
Catalog # NBP159553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ABCB8(ATP-binding cassette sub-family B (MDR/TAP) member 8) The peptide sequence was selected from the middle region of ABCB8. Peptide sequence EPVLFGTTIMENIRFGKLEASDEEVYTAAREANAHEFITSFPEGYNTVVG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABCB8; ATP-binding cassette sub-family B (MDR/TAP) member 8 EC 3.6.3 EST328128 M-ABC1mitochondrial ABC protein mitochondrial Mitochondrial ATP-binding cassette 1; ATP-binding cassette sub-family B member 8, mitochondrial; ATP-binding cassette, sub-family B (MDR/TAP), member 8; M-ABC1; mitochondrial ABC protein; Mitochondrial ATP-binding cassette 1; Mitochondrial potassium channel ATP-binding subunit; Mitochondrial sulfonylurea-receptor; MITOSUR
Gene Aliases: ABCB8; EST328128; M-ABC1; MABC1; MITOSUR
UniProt ID: (Human) Q9NUT2
Entrez Gene ID: (Human) 11194
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.