Novus Biologicals
Manufacturer Code:NBP303972
Catalog # NB094184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic Peptide of human ABCB7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC transporter 7 protein; ABC transporter 7 protein ABC7ATP-binding cassette sub-family B member 7 mitochondrial ASATATP-binding cassette 7 Atm1p ATP-binding cassette transporter 7 ATP-binding cassette sub-family B (MDR/TAP) member 7 EC 3.6.3 EC 3.6.3.42 EST140535; ATP-binding cassette sub-family B member 7; ATP-binding cassette sub-family B member 7, mitochondrial; ATP-binding cassette transporter 7; ATP-binding cassette, sub-family B (MDR/TAP), member 7
Gene Aliases: ABC7; ABCB7; ASAT; Atm1p; EST140535
UniProt ID: (Human) O75027
Entrez Gene ID: (Human) 22
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.