Novus Biologicals
Manufacturer Code:NBP292864100UL
Catalog # NB027593
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 503-753 of human ABCB7 (NP_004290.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC transporter 7 protein; ABC transporter 7 protein ABC7ATP-binding cassette sub-family B member 7 mitochondrial ASATATP-binding cassette 7 Atm1p ATP-binding cassette transporter 7 ATP-binding cassette sub-family B (MDR/TAP) member 7 EC 3.6.3 EC 3.6.3.42 EST140535; ATP-binding cassette sub-family B member 7; ATP-binding cassette sub-family B member 7, mitochondrial; ATP-binding cassette transporter 7; ATP-binding cassette, sub-family B (MDR/TAP), member 7
Gene Aliases: ABC7; ABCB7; ASAT; Atm1p; EST140535
UniProt ID: (Human) O75027
Entrez Gene ID: (Human) 22
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.