Proteintech
Manufacturer Code:111581AP150UL
Catalog # 50-172-6924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ABCB7 Fusion Protein Ag1544 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC transporter 7 protein; ABC transporter 7 protein ABC7 ABCB7 ASAT Atm1p EST140535; ATP-binding cassette 7; ATP-binding cassette sub-family B member 7; ATP-binding cassette sub-family B member 7, mitochondrial; ATP-binding cassette transporter 7; ATP-binding cassette, sub-family B (MDR/TAP), member 7; ATP-binding cassette, sub-family B, member 7, mitochondrial
Gene Aliases: AA517758; ABC7; ABCB7; ASAT; Atm1p; AU019072; EST140535
UniProt ID: (Human) O75027, (Mouse) Q61102, (Rat) Q704E8
Entrez Gene ID: (Human) 22, (Mouse) 11306, (Rat) 302395
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.