Novus Biologicals
Manufacturer Code:NBP258327
Catalog # NBP258327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:YYRMIQTNFIDMENMFDLLKEETEVKDLPGAGPLRFQKGRIEFENVHFSYADGRETLQD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC ATP-binding cassette sub-family B (MDR/TAP) member 6 EC 3.6.3 EST45597 FLJ22414 mitochondrial Mitochondrial ABC transporter 3 MTABC3ABC14 P-glycoprotein-related protein PRP UMAT; ATP-binding cassette half-transporter; ATP-binding cassette sub-family B member 6, mitochondrial; ATP-binding cassette, sub-family B (MDR/TAP), member 6 (Langereis blood group); Mitochondrial ABC transporter 3; Mt-ABC transporter 3; P-glycoprotein-related protein; Ubiquitously-expressed mammalian ABC half transporter
Gene Aliases: ABC; ABC14; ABCB6; DUH3; LAN; MCOPCB7; MTABC3; PRP; PSHK2; UMAT
UniProt ID: (Human) Q9NP58
Entrez Gene ID: (Human) 10058
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.