Proteintech
Manufacturer Code:149961AP150UL
Catalog # 50-172-9897
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ABCB6 Fusion Protein Ag6982 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC ABC14 ABCB6 EST45597 Mt ABC transporter 3 MTABC3 P glycoprotein related protein PRP umat; ATP-binding cassette half-transporter; ATP-binding cassette sub-family B member 6, mitochondrial; ATP-binding cassette, sub-family B (MDR/TAP), member 6; ATP-binding cassette, sub-family B (MDR/TAP), member 6 (Langereis blood group); Mitochondrial ABC transporter 3; Mt-ABC transporter 3; P-glycoprotein-related protein; Ubiquitously-expressed mammalian ABC half transporter
Gene Aliases: 1200005B17Rik; ABC; ABC14; ABCB6; DUH3; LAN; MCOPCB7; MTABC3; PRP; PSHK2; UMAT
UniProt ID: (Human) Q9NP58, (Rat) O70595, (Mouse) Q9DC29
Entrez Gene ID: (Human) 10058, (Rat) 140669, (Mouse) 74104
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.