Invitrogen
Manufacturer Code:PA518844
Catalog # PIPA518844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QTQHRNTSKKAQ ) corresponding to the internal amino acids of ABCB5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ABCB5 P-gp; ATP-binding cassette protein; ATP-binding cassette sub-family B (MDR/TAP) member 5 ABCB5alpha ABCB5beta EST422562 ATP-binding cassette sub-family B member 5N: P-glycoprotein ABCB5; ATP-binding cassette sub-family B member 5; ATP-binding cassette, sub-family B (MDR/TAP), member 5; P-glycoprotein ABCB5
Gene Aliases: ABCB5; ABCB5alpha; ABCB5beta; EST422562
UniProt ID: (Human) Q2M3G0
Entrez Gene ID: (Human) 340273
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.