Invitrogen
Manufacturer Code:PA5114801
Catalog # PIPA5114801
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ABCB5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9230106F14Rik Abcb5 ABCB5 P-gp ABCB5alpha ABCB5beta ATP binding cassette subfamily B member 5 ATP-binding cassette protein ATP-binding cassette sub-family B member 5 ATP-binding cassette subfamily B (MDR/TAP) member 5 ATP-binding cassette sub-family B (MDR/TAP) member 5 EST422562 P-glycoprotein ABCB5 RGD1566342; ABCB5 P-gp; ATP-binding cassette protein; ATP-binding cassette sub-family B member 5; ATP-binding cassette, sub-family B (MDR/TAP), member 5; ATP-binding cassette, subfamily B (MDR/TAP), member 5; P-glycoprotein ABCB5
Gene Aliases: 9230106F14Rik; ABCB5; ABCB5alpha; ABCB5beta; EST422562; RGD1566342
UniProt ID: (Human) Q2M3G0, (Mouse) B5X0E4
Entrez Gene ID: (Human) 340273, (Mouse) 77706, (Rat) 314537
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.