Novus Biologicals
Manufacturer Code:NBP177687AF647
Catalog # NBP177687Y
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal portion of the human ABCB5 protein (between residues 150-200) [UniProt Q2M3G0] |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABCB5 P-gp; ABCB5 P-gp ABCB5alpha ABCB5beta ATP-binding cassette protein ATP-binding cassette sub-family B member 5 ATP-binding cassette sub-family B (MDR/TAP) member 5 EC 3.6.3 EC 3.6.3.44 EST422562 P-glycoprotein ABCB5; ATP-binding cassette protein; ATP-binding cassette sub-family B member 5; ATP-binding cassette, sub-family B (MDR/TAP), member 5; P-glycoprotein ABCB5
Gene Aliases: ABCB5; ABCB5alpha; ABCB5beta; EST422562
UniProt ID: (Human) Q2M3G0
Entrez Gene ID: (Human) 340273
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.