Abnova
Manufacturer Code:MAB3415
Catalog # 89-111-406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | USal-hBSEP-McAb-1 |
Immunogen | A synthetic peptide corresponding to amino acids 705-718 of human ABCB11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of ABCB11.
For Research Use Only
Protein Aliases: ABC member 16, MDR/TAP subfamily; ABC16; ATP-binding cassette sub-family B member 11; ATP-binding cassette, sub-family B (MDR/TAP), member 11; Bile salt export pump; BSEP; PFIC-2; PFIC2; PGY4; progressive familial intrahepatic cholestasis 2; sister p-glycoprotein; SPGP
Gene Aliases: ABC16; ABCB11; BRIC2; BSEP; PFIC-2; PFIC2; PGY4; SPGP
UniProt ID: (Human) O95342
Entrez Gene ID: (Human) 8647
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.