Invitrogen
Manufacturer Code:PA513105
Catalog # PIPA513105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1038-1067 amino acids from the C-terminal region of human ABCB11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC member 16, MDR/TAP subfamily; ATP-binding cassette sub-family B member 11; ATP-binding cassette, sub-family B (MDR/TAP), member 11; ATP-binding cassette, sub-family B, member 11; Bile salt export pump; progressive familial intrahepatic cholestasis 2; Sister of P-glycoprotein; sister p-glycoprotein
Gene Aliases: ABC16; ABCB11; BRIC2; BSEP; Lith1; PFIC-2; PFIC2; PGY4; SPGP
UniProt ID: (Human) O95342, (Mouse) Q9QY30
Entrez Gene ID: (Human) 8647, (Mouse) 27413
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.