OriGene
Manufacturer Code:TA801025
Catalog # 50-167-8658
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B2 |
Immunogen | Human recombit protein fragment corresponding to amino acids 347-710 of human ABCB1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ABC20 CD243 CLCS GP170 MDR1 P-GP PGY1; ATP-binding cassette sub-family B member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1; ATP-dependent translocase ABCB1; CD243; colchicin sensitivity; doxorubicin resistance; Multidrug resistance protein 1; P-glycoprotein 1; Phospholipid transporter ABCB1
Gene Aliases: ABC20; ABCB1; CD243; CLCS; GP170; MDR1; P-GP; PGY1
UniProt ID: (Human) P08183
Entrez Gene ID: (Human) 5243
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.