Invitrogen
Manufacturer Code:PA536429
Catalog # PIPA536429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 1171-1220 of human ABCA8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC-type organic anion transporter ABCA8; ABC-type organic anion transporter ABCA8A; ATP-binding cassette sub-family A member 8; ATP-binding cassette sub-family A member 8 KIAA0822; ATP-binding cassette sub-family A member 8-A; ATP-binding cassette sub-family A member 8A; ATP-binding cassette, sub-family A (ABC1), member 8; ATP-binding cassette, sub-family A (ABC1), member 8b; ATP-binding cassette, subfamily A (ABC1), member 8
Gene Aliases: ABCA8; Abca8a; Abca8b; KIAA0822
UniProt ID: (Human) O94911, (Mouse) Q8K442
Entrez Gene ID: (Human) 10351, (Rat) 303637, (Mouse) 217258
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.