Novus Biologicals
Manufacturer Code:NBP120867
Catalog # NBP120867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KEQELGDFEEDFD corresponding to C-Terminus according to NP_009099.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC-type organic anion transporter ABCA8; ATP-binding cassette sub-family A member 8; ATP-binding cassette sub-family A member 8 ATP-binding cassette sub-family A (ABC1) member 8 EC 3.6.3 EC 3.6.3.41 KIAA0822MGC163152; ATP-binding cassette, sub-family A (ABC1), member 8
Gene Aliases: ABCA8; KIAA0822
UniProt ID: (Human) O94911
Entrez Gene ID: (Human) 10351
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.