Novus Biologicals
Manufacturer Code:NB400166
Catalog # NB400166
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 7A1-144 |
Immunogen | Mouse ABCA7 stably-transfected HeLa cells. [UniProt# Q91V24] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ABCA-SSN; ABCA-SSN ABCXATP-binding cassette sub-family A member 7 ATP-binding cassette sub-family A (ABC1) member 7 Autoantigen SS-N EC 2.2.1.1 EC 3.6.3 EC 3.6.3.41 FLJ40025 Macrophage ABC transporter; ATP-binding cassette sub-family A member 7; ATP-binding cassette, sub-family A (ABC1), member 7; Autoantigen SS-N; Macrophage ABC transporter; Phospholipid-transporting ATPase ABCA7
Gene Aliases: ABCA-SSN; ABCA7; ABCX; AD9
UniProt ID: (Human) Q8IZY2
Entrez Gene ID: (Human) 10347
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.