Invitrogen
Manufacturer Code:PA559624
Catalog # PIPA559624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ABCA7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC 1 antibody ABC Transporter 1 antibody ABC1 antibody ATP-binding Cassette 1 antibody ATP-binding Cassette Transporter 1 antibody CERP antibody; ABCA-SSN; ATP-binding cassette sub-family A member 7; ATP-binding cassette, sub-family A (ABC1), member 7; Autoantigen SS-N; Macrophage ABC transporter; Phospholipid-transporting ATPase ABCA7
Gene Aliases: ABCA-SSN; ABCA7; ABCX; AD9
UniProt ID: (Human) Q8IZY2
Entrez Gene ID: (Human) 10347
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.