Invitrogen
Manufacturer Code:PA5111558
Catalog # PIPA5111558
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human ABCA5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC13 ABCA5 ATP-binding cassette sub-family A member 5 B930033A02Rik DKFZp451F117 DKFZp779N2435 EST90625 FLJ16381 Kiaa1888 mKIAA1888; ATP-binding cassette A5; ATP-binding cassette sub-family A member 5; ATP-binding cassette, sub-family A (ABC1), member 5; Cholesterol transporter ABCA5
Gene Aliases: ABC13; ABCA5; EST90625; KIAA1888
UniProt ID: (Human) Q8WWZ7
Entrez Gene ID: (Human) 23461
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.