Invitrogen
Manufacturer Code:PA578689
Catalog # PIPA578689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of mouse ABCA4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-binding cassette 10; ATP-binding cassette sub-family A member 4; ATP-binding cassette, sub-family A (ABC1), member 4; Retinal-specific ATP-binding cassette transporter; Retinal-specific ATP-binding cassette transporter ATP-binding cassette sub-family A member 4 RIM ABC transporter RIM protein RmP Abca4 Abcr; Retinal-specific phospholipid-transporting ATPase ABCA4; RIM ABC transporter; RIM protein
Gene Aliases: Abc10; Abca4; ABCR; AW050280; D430003I15Rik; RmP
UniProt ID: (Mouse) O35600
Entrez Gene ID: (Mouse) 11304, (Rat) 310836
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.