Novus Biologicals
Manufacturer Code:NBP29260120UL
Catalog # NB027073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 110-300 of human ABCA4 (NP_000341.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC10 ABCRRMP ARMD2DKFZp781N1972 ATP binding cassette transporter ATP-binding cassette sub-family A member 4 ATP-binding cassette transporter retinal-specific ATP-binding cassette sub-family A (ABC1) member 4 ATP-binding transporter retina-specific CORD3 EC 3.6.3 FFMFLJ17534 photoreceptor rim protein retinal-specific ATP-binding cassette transporter retina-specific ABC transporter RIM ABC transporter RIM protein RmP RP19 Stargardt disease protein STGD STGD1; ATP binding cassette transporter; ATP-binding cassette sub-family A member 4; ATP-binding cassette transporter, retinal-specific; ATP-binding cassette, sub-family A (ABC1), member 4; ATP-binding transporter, retina-specific; photoreceptor rim protein; retina-specific ABC transporter; Retinal-specific ATP-binding cassette transporter; Retinal-specific phospholipid-transporting ATPase ABCA4; RIM ABC transporter; RIM protein; Stargardt disease protein
Gene Aliases: ABC10; ABCA4; ABCR; ARMD2; CORD3; FFM; RMP; RP19; STGD; STGD1
UniProt ID: (Human) P78363
Entrez Gene ID: (Human) 24
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.