Invitrogen
Manufacturer Code:PA568105
Catalog # PIPA568105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human ABCA3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 150 Kda mature form; ABC transporter 3; ABC-C transporter; ABC3; ATP-binding cassette 3; ATP-binding cassette sub-family A member 3; ATP-binding cassette transporter 3; ATP-binding cassette, sub-family A (ABC1), member 3; Phospholipid-transporting ATPase ABCA3; Xenobiotic-transporting ATPase ABCA3
Gene Aliases: 1810036E22Rik; ABC-C; ABC3; ABCA3; EST111653; LBM180; SMDP3
UniProt ID: (Human) Q99758, (Mouse) Q8R420
Entrez Gene ID: (Human) 21, (Mouse) 27410
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.