Novus Biologicals
Manufacturer Code:NBP292667100UL
Catalog # NB027203
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 43-203 of human ABCA3 (NP_001080.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 150 Kda mature form; ABC transporter 3; ABC transporter 3 ABC3MGC72201 ABC-C ABC-C transporter ATP-binding cassette 3 ATP-binding cassette sub-family A member 3 ATP-binding cassette transporter 3 ATP-binding cassette sub-family A (ABC1) member 3 EC 3.6.3 EC 3.6.3.17 EST111653 LBM180 MGC166979 SMDP3; ABC-C transporter; ATP-binding cassette 3; ATP-binding cassette sub-family A member 3; ATP-binding cassette transporter 3; ATP-binding cassette, sub-family A (ABC1), member 3; Phospholipid-transporting ATPase ABCA3; Xenobiotic-transporting ATPase ABCA3
Gene Aliases: ABC-C; ABC3; ABCA3; EST111653; LBM180; SMDP3
UniProt ID: (Human) Q99758
Entrez Gene ID: (Human) 21
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.